Merck
  • Colistin methanesulfonate and colistin pharmacokinetics in critically ill patients receiving continuous venovenous hemodiafiltration.
Colistin methanesulfonate and colistin pharmacokinetics in critically ill patients receiving continuous venovenous hemodiafiltration.
Antimicrobial agents and chemotherapy (2012-11-14)
Matti Karvanen, Diamantis Plachouras, Lena E Friberg, Elisabeth Paramythiotou, Evangelos Papadomichelakis, Ilias Karaiskos, Iraklis Tsangaris, Apostolos Armaganidis, Otto Cars, Helen Giamarellou
ABSTRACT
This report describes the pharmacokinetics of colistin methanesulfonate (CMS) and colistin in five intensive care unit patients receiving continuous venovenous hemodiafiltration. For CMS, the mean maximum concentration of drug in plasma (C(max)) after the fourth dose was 6.92 mg/liter and total clearance (CL) 8.23 liters/h. For colistin, the mean concentration was 0.92 mg/liter and CL/metabolized fraction (f(m)) 18.91 liters/h. Colistin concentrations were below the current MIC breakpoints, and the area under the concentration-time curve for the free, unbound fraction of the drug over 24 h in the steady state divided by the MIC (fAUC/MIC) was lower than recommended, suggesting that a dosage regimen of 160 mg CMS every 8 h (q8h) is inadequate.
Sigma-Aldrich
Colistin sodium methanesulfonate, ~11,500 U/mg
Sigma-Aldrich
Colistin sodium methanesulfonate, from Bacillus colistinus
Colistimethate sodium, European Pharmacopoeia (EP) Reference Standard