Merck
NMR chemical shift in an electronic state with arbitrary degeneracy.
Physical review letters (2012-09-26)
Willem Van den Heuvel, Alessandro Soncini
ABSTRACT
We present a theory of nuclear magnetic resonance shielding tensors for electronic states with arbitrary degeneracy. The shieldings are here expressed in terms of generalized Zeeman (g((k))) and hyperfine (A((k))) tensors, of all ranks k allowed by the size of degeneracy. Contrary to recent proposals [T. O. Pennanen and J. Vaara, Phys. Rev. Lett. 100, 133002 (2008)], our theory is valid in the strong spin-orbit coupling limit. Ab initio calculations for the fourfold degenerate Γ(8) ground state of lanthanide-doped fluorite crystals CaF(2):Ln (Ln=Pr(2+), Nd(3+), Sm(3+), and Dy(3+)) show that previously neglected contributions can account for more than 50% of the paramagnetic shift.
Sigma-Aldrich
Calcium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium fluoride, random crystals, optical grade, 99.99% trace metals basis