Merck
  • A genome-wide single-nucleotide polymorphism-array can improve the prognostic stratification of the core binding factor acute myeloid leukemia.
A genome-wide single-nucleotide polymorphism-array can improve the prognostic stratification of the core binding factor acute myeloid leukemia.
American journal of hematology (2012-08-14)
Jungwon Huh, Hee-Je Kim, Chul Won Jung, Hee-Jin Kim, Sun-Hee Kim, Yeo-Kyeoung Kim, Hyeoung-Joon Kim, Myung Geun Shin, Joon Ho Moon, Sang Kyun Sohn, Sung Hyun Kim, Won Sik Lee, Jong Ho Won, Yeung Chul Mun, Hawk Kim, Jinny Park, Woo Sung Min, Dong Hwan Dennis Kim
ABSTRACT
Core binding factor (CBF) AML with the D816 C-KIT gene mutation demonstrate inferior treatment outcomes. However, the remaining cases without the D816 C-KIT mutation imply a requirement of more sophisticated dissection of the patients according to their prognosis. In this study, we analyzed the prognostic value of a single nucleotide polymorphism array (SNP-A) based karyotyping combined with metaphase cytogenetics (MC) to facilitate further stratification of CBF AML patients. A total of 98 CBF AML patients were included and genome-wide Human SNP 6.0 Arrays (Affymetrix) were performed using marrow samples taken at diagnosis. Overall, 40 abnormal lesions were identified in 25 patients (26%). Survival of the patients with the abnormal lesion(s) detected by SNP-A and/or MC was worse than those without lesions in terms of the 2-year overall survival (OS; 57.5% vs. 76.4%, P = 0.028), event-free (EFS; 45.7% vs. 66.2%, P = 0.072), and leukemia-free survival (LFS; 49.0% vs. 77.4%, P = 0.015), specially in the subgroup with inv(16)/t(16;16) (40.9% vs. 80.2% OS, P = 0.040) and in the subgroup without the D816 C-KIT mutation (61.6% vs. 82.7% OS, P = 0.038). Multivariate analysis confirmed the prognostic impact of the abnormal SNP-A and/or MC lesion on EFS (HR 2.011, P = 0.047), and LFS (HR 3.231, P = 0.005) in the overall CBF AML. This study suggests that the combined use of SNP-A with MC in the CBF AML can provide important prognostic value, especially in the inv(16)/t(16;16) subgroup or in the patients without the D816 C-KIT mutation.
Sigma-Aldrich
Idarubicin hydrochloride, solid