Merck
  • Melanosome-iron interactions within retinal pigment epithelium-derived cells.
Melanosome-iron interactions within retinal pigment epithelium-derived cells.
Pigment cell & melanoma research (2012-08-14)
Patrycja Kaczara, Mariusz Zaręba, Anja Herrnreiter, Christine M B Skumatz, Andrzej Ządło, Tadeusz Sarna, Janice M Burke
ABSTRACT
Melanosomes were recently shown to protect ARPE-19 cells, a human retinal pigment epithelium (RPE) cell line, against oxidative stress induced by hydrogen peroxide. One postulated mechanism of antioxidant action of melanin is its ability to bind metal ions. The aim here was to determine whether melanosomes are competent to bind iron within living cells, exhibiting a property previously shown only in model systems. The outcomes indicate retention of prebound iron and accumulation of iron by granules after iron delivery to cells via the culture medium, as determined by both colorimetric and electron spin resonance analyses for bound-to-melanosome iron. Manipulation of iron content did not affect the pigment's ability to protect cells against H(2) O(2) , but the function of pigment granules within RPE cells should be extended beyond a role in light irradiation to include participation in iron homeostasis.
Sigma-Aldrich
Apoferritin from equine spleen
Sigma-Aldrich
Apoferritin from equine spleen, 0.2 μm filtered