Merck
Flotation of marine microalgae: effect of algal hydrophobicity.
Bioresource technology (2012-08-04)
Sourabh Garg, Yan Li, Liguang Wang, Peer M Schenk
ABSTRACT
This study aims to understand the underlying reasons for the poor flotation response of marine microalgae. The flotation performance and hydrophobicity of a freshwater microalga (Chlorella sp. BR2) were compared to those of a marine microalga (Tetraselmis sp. M8) at different salinities in the presence of a cationic collector, tetradecyl trimethylammonium bromide. It was found that microalgal hydrophobicity played a more important role than salinity in determining the flotation performance.
Product Description
Supelco
Myristyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Trimethyl-tetradecylammonium chloride, ≥98.0% (AT)
Sigma-Aldrich
Myristyltrimethylammonium bromide, 98% (AT)
Sigma-Aldrich
Myristyltrimethylammonium bromide, ≥99%