Merck
  • Effect of preservatives on Shiga toxigenic phages and Shiga toxin of Escherichia coli O157:H7.
Effect of preservatives on Shiga toxigenic phages and Shiga toxin of Escherichia coli O157:H7.
Journal of food protection (2012-05-09)
Tomás Subils, Virginia Aquili, Guillermo Ebner, Claudia Balagué
ABSTRACT
Toxin synthesis by Shiga toxin-producing Escherichia coli (STEC) appears to be coregulated through the induction of the integrated bacteriophages that encode the toxin genes. These phages might be the principal means for the dissemination and release of Shiga toxins. We evaluated the effect of three common food preservatives, potassium sorbate, sodium benzoate, and sodium propionate, on the propagation of the phages and Shiga toxins. We tested each preservative at four concentrations, 1, 1.25, 2.5, and 5 mg/ml, both on free phages and on lysogenic phages in bacteria. We also evaluated the expression of a lambdoid phage, which was exposed to increasing concentrations of preservatives, by measuring β-galactosidase activity from SPC105, a transductant strain. Furthermore, we tested the effect of the preservatives on cytotoxigenic activity of Shiga toxin on Vero cells. We detected an increase of the inhibitory effect of the phage lytic activity, both in lysogenic and free phages, as the preservative concentration increased. However, the inhibition was higher on the lysogenic phages release than on free phages. Sodium benzoate and potassium sorbate were about equal at inhibiting phages; they were more effective than sodium propionate. A significant decrease of lacZ expression, encoded in a lambda phage, was observed. We also found a reduction in Shiga toxin titer caused by exposure of E. coli O157:H7 to 5 mg/ml sodium benzoate or potassium sorbate. These results imply that these three preservatives, used to inhibit microbial spoilage of foods, also act to inhibit lytic activity and dispersion of a phage carrying the gene encoding powerful Shiga cytotoxins. Also notable was the inactivation of Shiga toxin activity, although this effect was detected using concentrations of preservatives greater than those allowed by the Argentine Food Code.
Product Description
Supelco
Sodium benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium benzoate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
Sodium benzoate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium benzoate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium benzoate, purum p.a., ≥99.0% (NT)