Merck
  • Type I interferons impair BDNF-induced cell signaling and neurotrophic activity in differentiated human SH-SY5Y neuroblastoma cells and mouse primary cortical neurons.
Type I interferons impair BDNF-induced cell signaling and neurotrophic activity in differentiated human SH-SY5Y neuroblastoma cells and mouse primary cortical neurons.
Journal of neurochemistry (2012-04-27)
Simona Dedoni, Maria C Olianas, Angela Ingianni, Pierluigi Onali
ABSTRACT
Type I interferons (IFNs) have been shown to act on neurons and to cause neuronal damage through mechanisms not completely defined. Here, we investigated the effects of type I IFNs on brain-derived neurotrophic factor (BDNF)-induced TrkB receptor signaling and neurotrophic activity. In retinoic acid-treated human SH-SY5Y neuroblastoma cells and mouse primary cortical neurons, long-term exposure to IFNs curtailed BDNF-induced activation of phosphatidylinositol 3-kinase, phospholipase Cγ and extracellular-regulated kinases 1 and 2 signaling. Moreover, IFN-β inhibited BDNF-induced cell survival, neurite outgrowth, and expression of neuronal markers, such as neurofilament proteins, growth-associated protein-43 and glutamate α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptor subunit GluR1. The IFN inhibitory effects were associated with down-regulation of TrkB and inhibition of TrkB autophosphorylation. In SH-SY5Y cells, blockade of either Janus kinase with pyridone 6 or signal transducer and activator of transcription (STAT) 1 with siRNA transfection attenuated IFN-β-induced TrkB down-regulation. Quantitative real time RT-PCR indicated that IFN-β significantly reduced TrkB mRNA levels. Moreover, blockade of protein kinase R counteracted IFN-β-induced inhibition of TrkB expression and signaling. These data indicate that in neuronal cells IFNs negatively regulate BDNF signaling and neurotrophic activity through inhibition of TrkB activation and Janus kinase/Signal transducer and activator of transcription-dependent down-regulation of TrkB.
Sigma-Aldrich
PKR Inhibitor, The PKR Inhibitor, also referenced under CAS 608512-97-6, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Oxotremorine M, solid