Merck
  • Ultrathin nanosheets of Li2MSiO4 (M = Fe, Mn) as high-capacity Li-ion battery electrode.
Ultrathin nanosheets of Li2MSiO4 (M = Fe, Mn) as high-capacity Li-ion battery electrode.
Nano letters (2012-02-16)
Dinesh Rangappa, Kempaiah Devaraju Murukanahally, Takaaki Tomai, Atsushi Unemoto, Itaru Honma
ABSTRACT
Novel ultrathin Li(2)MnSiO(4) nanosheets have been prepared in a rapid one pot supercritical fluid synthesis method. Nanosheets structured cathode material exhibits a discharge capacity of ~340 mAh/g at 45 ± 5 °C. This result shows two lithium extraction/insertion performances with good cycle ability without any structural instability up to 20 cycles. The two-dimensional nanosheets structure enables us to overcome structural instability problem in the lithium metal silicate based cathode materials and allows successful insertion/extraction of two complete lithium ions.
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)