Merck
  • Lipase-coated K2SO4 micro-crystals: preparation, characterization, and application in biodiesel production using various oil feedstocks.
Lipase-coated K2SO4 micro-crystals: preparation, characterization, and application in biodiesel production using various oil feedstocks.
Bioresource technology (2012-02-15)
Jingjing Zheng, Li Xu, Yun Liu, Xiaoyan Zhang, Yunjun Yan
ABSTRACT
This study investigated the preparation and characteristics of protein-coated microcrystals (PCMCs) from Pseudomonas cepacia lipase (PS) and K(2)SO(4), and their application in biodiesel synthesis, via single factorial experiments and response surface methodology (RSM), the optimized PCMC-PS exhibited high activity and stability; the optimal temperature was 60°C (which gave 99.83% conversion), although fairly high activity was exhibited after incubation at different temperatures (25-70°C). The organic solvents stability of the PCMC-PS was improved, and it significantly reduced ethanol toxicity. Circular dichroism (CD) analysis revealed the correlation between the conformation and the enzyme activity. The morphology of the PCMC-PS was also confirmed via scanning electron microscopy (SEM). When catalyzed by PCMC-PS, above 83% biodiesel yield was obtained for most of the seven oils tested. The PCMC-PS (washed with hexane) activity remained relatively stable after eight batch reactions, with only a 15.73% reduction in the conversion (from 99.02% to 83.29%).
Sigma-Aldrich
Potassium sulfate, suitable for plant cell culture
Sigma-Aldrich
Potassium bisulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium sulfate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Potassium sulfate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Potassium sulfate, meets analytical specification of Ph. Eur., 99-101%
Sigma-Aldrich
Potassium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium sulfate, ACS reagent, ≥99.0%, powder or crystals
Sigma-Aldrich
Potassium bisulfate, reagent grade