Merck
  • Complete genome sequence of the chloromethane-degrading Hyphomicrobium sp. strain MC1.
Complete genome sequence of the chloromethane-degrading Hyphomicrobium sp. strain MC1.
Journal of bacteriology (2011-08-27)
Stéphane Vuilleumier, Thierry Nadalig, Muhammad Farhan Ul Haque, Ghislaine Magdelenat, Aurélie Lajus, Sandro Roselli, Emilie E L Muller, Christelle Gruffaz, Valérie Barbe, Claudine Médigue, Françoise Bringel
ABSTRACT
Hyphomicrobium sp. strain MC1 is an aerobic methylotroph originally isolated from industrial sewage. This prosthecate bacterium was the first strain reported to grow with chloromethane as the sole carbon and energy source. Its genome, consisting of a single 4.76-Mb chromosome, is the first for a chloromethane-degrading bacterium to be formally reported.
Product Description
Supelco
Chloromethane solution, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
Chloromethane solution, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Chloromethane solution, 1.0 M in diethyl ether
Sigma-Aldrich
Chloromethane, ≥99.5%