Merck
  • Fast high-resolution 2D correlation spectroscopy in inhomogeneous fields via Hadamard intermolecular multiple quantum coherences technique.
Fast high-resolution 2D correlation spectroscopy in inhomogeneous fields via Hadamard intermolecular multiple quantum coherences technique.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-06-15)
Congbo Cai, Fenglian Gao, Shuhui Cai, Yuqing Huang, Zhong Chen
ABSTRACT
Recently, a method based on intermolecular multiple quantum coherences (iMQCs) has been proposed to obtain high-resolution 2D COSY spectra in inhomogeneous fields via 3D acquisitions. However, the very long acquisition time prevents its practical application. To overcome this shortage, the Hadamard technique was applied for the iMQC method in this paper. For the new pulse sequence, the direct frequency-domain excitation is used in the first indirect detection dimension, so the 3D acquisition was replaced by an array of 2D acquisitions. The acquisition time can be reduced to 10 min. The resulting spectra retain useful structural information including chemical shifts and multiplet patterns of J coupling even when the inhomogeneous line broadening leads to overlap of neighboring diagonal resonances in the conventional COSY spectrum. The experimental results are consistent with the theoretical predictions and computer simulations. The new sequence may provide a time-efficient way for the studies of chemical solution in inhomogeneous fields.
Sigma-Aldrich
1-Bromobutane, ReagentPlus®, 99%
Supelco
1-Bromobutane, analytical standard