Merck
  • Concise synthesis of Valerena-4,7(11)-diene, a highly active sedative, from valerenic acid.
Concise synthesis of Valerena-4,7(11)-diene, a highly active sedative, from valerenic acid.
Bioscience, biotechnology, and biochemistry (2010-09-14)
Takashi Kitayama, Genki Kawabata, Michiho Ito
ABSTRACT
A concise synthesis of valerena-4,7(11)-diene with potent sedative activity was achieved in three steps involving, reduction of carboxylic acid, bromination of the resulting alcohol, and reduction of the bromide from valerenic acid in a 63% total yield. This synthetic method makes it possible to provide further materials for biological testing to realize comprehensive SAR studies.
Product Description
Supelco
Valerenic acid, analytical standard
Valerenic acid, primary reference standard