Merck
  • CuCl/CCl(4)-promoted convenient synthesis of sulfonyl amidines from tertiary amines and sulfonyl azides.
CuCl/CCl(4)-promoted convenient synthesis of sulfonyl amidines from tertiary amines and sulfonyl azides.
Organic letters (2010-02-04)
Xiaoliang Xu, Zhichuang Ge, Dongping Cheng, Lei Ma, Chunshan Lu, Qunfeng Zhang, Nan Yao, Xiaonian Li
ABSTRACT
Promoted by CuCl/CCl(4), a variety of sulfonyl azides and tertiary amines were successfully coupled to give sulfonyl amidine derivatives in good to excellent yields. A possible mechanism for this reaction is discussed.
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%