Merck
  • Dispersive liquid-liquid-liquid microextraction combined with liquid chromatography for the determination of chlorophenoxy acid herbicides in aqueous samples.
Dispersive liquid-liquid-liquid microextraction combined with liquid chromatography for the determination of chlorophenoxy acid herbicides in aqueous samples.
Journal of chromatography. A (2009-10-07)
Wan-Chun Tsai, Shang-Da Huang
ABSTRACT
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In this study, 25microL of 1,1,2,2-tetrachloroethane was added to the sample solution and the targeted analytes were extracted from the donor phase by manually shaking for 90s. The organic phase was separated from the donor phase by centrifugation and was transferred into an insert. Acceptor phase was added to this insert. The analytes were then back-extracted into the acceptor phase by mixing the organic and acceptor phases by pumping those two solutions with a syringe plunger. After centrifugation, the organic phase was settled and removed with a microsyringe. The acceptor phase was injected into the UPLC system by auto sampler. Fine droplets were formed by shaking and pumping with the syringe plunger in DLLLME. The large interfacial area provided good extraction efficiency and shortened the extraction time needed. Conventional LLLME requires an extraction time of 40-60min; an extraction time of approximately 2min is sufficient with DLLLME. The DLLLME technique shows good linearity (r(2)>or=0.999), good repeatability (RSD: 4.0-12.2% for tap water; 5.7-8.5% for river water) and high sensitivity (LODs: 0.10-0.60microg/L for tap water; 0.11-0.95microg/L for river water).
Sigma-Aldrich
1,1,2,2-Tetrachloroethane, reagent grade, ≥98%
Sigma-Aldrich
1,1,2,2-Tetrachloroethane, JIS special grade, ≥97.0%
Supelco
1,1,2,2-Tetrachloroethane, analytical standard