Merck
Ceric ammonium nitrate promoted oxidation of oxazoles.
Organic letters (2006-11-17)
David A Evans, Pavel Nagorny, Risheng Xu
ABSTRACT
The ceric ammonium nitrate promoted oxidations of 4,5-diphenyloxazoles and oxazoles with various substitution patterns have been investigated. This transformation results in the formation of the corresponding imide in good yield and tolerates a wide variety of functional groups and substituents on the oxazole moiety. [reaction: see text].
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, JIS special grade, ≥95.0%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)