Merck
  • Quantitative structure-activity relationship analyses of antioxidant and free radical scavenging activities for hydroxybenzalacetones.
Quantitative structure-activity relationship analyses of antioxidant and free radical scavenging activities for hydroxybenzalacetones.
Bioorganic & medicinal chemistry letters (2004-10-16)
Chisako Yamagami, Noriko Motohashi, Tatsuhiko Emoto, Akira Hamasaki, Takao Tanahashi, Naotaka Nagakura, Yoshito Takeuchi
ABSTRACT
Antioxidant activities for a series of hydroxybenzalacetones, OH-BZ, were evaluated by measuring inhibitory potencies of OH-BZ against lipid peroxidation induced by t-BuOOH or gamma-irradiation. Their quantitative structure-activity relationship (QSAR) studies indicated that the activities are mainly governed by electronic and steric factors. To rationalize these results, we also performed QSAR analyses for DPPH radical scavenging activities of OH-BZ, which indicated that antioxidant and radical scavenging activities could be expressed by the same physicochemical parameters but the hydrogen bonding behavior of phenolic OH varies with the reaction medium.
Sigma-Aldrich
trans-4-Phenyl-3-buten-2-one, ≥99%
Sigma-Aldrich
4-Phenyl-3-buten-2-one, 99%
Sigma-Aldrich
Benzylideneacetone, ≥98%, FG