Merck
  • Protective effect of green tea polyphenol (-)-epigallocatechin gallate and other antioxidants on lipid peroxidation in gerbil brain homogenates.
Protective effect of green tea polyphenol (-)-epigallocatechin gallate and other antioxidants on lipid peroxidation in gerbil brain homogenates.
Phytotherapy research : PTR (2003-04-03)
Seong-Ryong Lee, Ki-Jo Im, Seong-Il Suh, Jung-Gil Jung
ABSTRACT
The aim of this study was to compare the protective effects of green tea polyphenol (-)-epigallocatechin gallate (EGCG) and other well-known antioxidants on the lipid peroxidation in gerbil brain homogenates. Oxidative stress was induced by H2O2 (10 mM) or ferrous ammonium sulfate (5 microM) and lipid peroxidation was studied. Hydrogen peroxide and ferrous ions are capable of oxidizing a wide range of substrates and causing biological damage. The reaction, referred to as the Fenton process, is complex and can generate both hydroxyl radicals and higher oxidation states of the iron. Thiobarbituric acid-reactive substances (TBA-RS) were used as a marker of lipid peroxidation. EGCG, trolox, lipoic acid, and melatonin reduced H2O2- or ferrous ion-induced lipid peroxidation in a concentration-dependent manner. In reducing the H2O2-induced lipid peroxidation, IC50 values of antioxidants were as follows: EGCG (0.66 microM), trolox (37.08 microM), lipoic acid (7.88 mM), and melatonin (19.11 mM). In reducing the ferrous ion-induced lipid peroxidation, IC50 values of antioxidants were as follows: EGCG (3.32 microM), trolox (75.65 microM), lipoic acid (7.63 mM), and melatonin (15.48 mM). Under the in vitro conditions of this experiment, EGCG was the most potent antioxidant in inhibiting H2O2 or ferrous ion-induced lipid peroxidation in the gerbil brain homogenates.
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, 99.997% trace metals basis
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ReagentPlus®, ≥98%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ACS reagent, 99%