Merck
  • Intestinal and hepatic microsomal metabolism of testosterone and progesterone by a 3 alpha-hydroxysteroid dehydrogenase to the 3 alpha-hydroxy derivatives in the channel catfish, Ictalurus punctatus.
Intestinal and hepatic microsomal metabolism of testosterone and progesterone by a 3 alpha-hydroxysteroid dehydrogenase to the 3 alpha-hydroxy derivatives in the channel catfish, Ictalurus punctatus.
The Journal of steroid biochemistry and molecular biology (2003-02-19)
Zhen Lou, Jodie V Johnson, Margaret O James
ABSTRACT
Intestinal or hepatic microsomes from channel catfish converted [4-14C]-testosterone to three major metabolites: 6 beta-hydroxytestosterone, androstenedione and a third metabolite. Formation of the unknown metabolite required NADPH as cofactor. When incubated with 200 microM testosterone, the rate of formation of the unknown metabolite was 265+/-158 pmol/(min mg) protein (mean+/-S.D.) in microsomes from the proximal intestine, 515+/-93 pmol/(min mg) protein in distal intestine and 226+/-42 pmol/(min mg) protein in hepatic microsomes. Comparison of the chromatographic and spectral properties of the unknown metabolite with those of authentic testosterone derivatives showed that this metabolite corresponded to 4-androstene-3 alpha,17 beta-diol. No 3 alpha-reduced metabolite was formed in incubations of testosterone with catfish intestinal cytosol. Testosterone was reduced to 5 alpha-dihydrotestosterone primarily in the cytosolic fraction and not in microsomes. Incubation of progesterone with intestinal microsomes resulted in the formation of a metabolite with properties similar to that of the 3 alpha-reduced testosterone, and this metabolite was identified by co-chromatography with authentic standard as 3 alpha-reduced progesterone. Thus, 3 alpha-hydroxysteroid dehydrogenase is an important pathway in intestinal microsomes of the channel catfish.
Sigma-Aldrich
6β-Hydroxytestosterone, ≥97% (HPLC)
Supelco
6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®