Merck
  • Neuronal DSCAM regulates the peri-synaptic localization of GLAST in Bergmann glia for functional synapse formation.
Neuronal DSCAM regulates the peri-synaptic localization of GLAST in Bergmann glia for functional synapse formation.
Nature communications (2024-02-02)
Ken-Ichi Dewa, Nariko Arimura, Wataru Kakegawa, Masayuki Itoh, Toma Adachi, Satoshi Miyashita, Yukiko U Inoue, Kento Hizawa, Kei Hori, Natsumi Honjoya, Haruya Yagishita, Shinichiro Taya, Taisuke Miyazaki, Chika Usui, Shoji Tatsumoto, Akiko Tsuzuki, Hirotomo Uetake, Kazuhisa Sakai, Kazuhiro Yamakawa, Takuya Sasaki, Jun Nagai, Yoshiya Kawaguchi, Masaki Sone, Takayoshi Inoue, Yasuhiro Go, Noritaka Ichinohe, Kozo Kaibuchi, Masahiko Watanabe, Schuichi Koizumi, Michisuke Yuzaki, Mikio Hoshino
ABSTRACT
In the central nervous system, astrocytes enable appropriate synapse function through glutamate clearance from the synaptic cleft; however, it remains unclear how astrocytic glutamate transporters function at peri-synaptic contact. Here, we report that Down syndrome cell adhesion molecule (DSCAM) in Purkinje cells controls synapse formation and function in the developing cerebellum. Dscam-mutant mice show defects in CF synapse translocation as is observed in loss of function mutations in the astrocytic glutamate transporter GLAST expressed in Bergmann glia. These mice show impaired glutamate clearance and the delocalization of GLAST away from the cleft of parallel fibre (PF) synapse. GLAST complexes with the extracellular domain of DSCAM. Riluzole, as an activator of GLAST-mediated uptake, rescues the proximal impairment in CF synapse formation in Purkinje cell-selective Dscam-deficient mice. DSCAM is required for motor learning, but not gross motor coordination. In conclusion, the intercellular association of synaptic and astrocyte proteins is important for synapse formation and function in neural transmission.
Sigma-Aldrich
Anti-DSCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-MARCKS antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Riluzole, A cell-permeable benzothiazole compound that potently inhibits glutamate release and blocks Na+ channels.
Sigma-Aldrich
Anti-FLAG Tag antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-phospho-MARCKS (Ser152/156) Antibody, from rabbit, purified by affinity chromatography