Merck
  • Galectin-9 mediates neutrophil capture and adhesion in a CD44 and β2 integrin-dependent manner.
Galectin-9 mediates neutrophil capture and adhesion in a CD44 and β2 integrin-dependent manner.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2021-12-01)
Asif J Iqbal, Franziska Krautter, Isobel A Blacksell, Rachael D Wright, Shani N Austin-Williams, Mathieu-Benoit Voisin, Mohammed T Hussain, Hannah L Law, Toshiro Niki, Mitsuomi Hirashima, Michele Bombardieri, Costantino Pitzalis, Alok Tiwari, Gerard B Nash, Lucy V Norling, Dianne Cooper
ABSTRACT
Neutrophil trafficking is a key component of the inflammatory response. Here, we have investigated the role of the immunomodulatory lectin Galectin-9 (Gal-9) on neutrophil recruitment. Our data indicate that Gal-9 is upregulated in the inflamed vasculature of RA synovial biopsies and report the release of Gal-9 into the extracellular environment following endothelial cell activation. siRNA knockdown of endothelial Gal-9 resulted in reduced neutrophil adhesion and neutrophil recruitment was significantly reduced in Gal-9 knockout mice in a model of zymosan-induced peritonitis. We also provide evidence for Gal-9 binding sites on human neutrophils; Gal-9 binding induced neutrophil activation (increased expression of β2 integrins and reduced expression of CD62L). Intra-vital microscopy confirmed a pro-recruitment role for Gal-9, with increased numbers of transmigrated neutrophils following Gal-9 administration. We studied the role of both soluble and immobilized Gal-9 on human neutrophil recruitment. Soluble Gal-9 significantly strengthened the interaction between neutrophils and the endothelium and inhibited neutrophil crawling on ICAM-1. When immobilized, Gal-9 functioned as an adhesion molecule and captured neutrophils from the flow. Neutrophils adherent to Gal-9 exhibited a spread/activated phenotype that was inhibited by CD18 and CD44 neutralizing antibodies, suggesting a role for these molecules in the pro-adhesive effects of Gal-9. Our data indicate that Gal-9 is expressed and released by the activated endothelium and functions both in soluble form and when immobilized as a neutrophil adhesion molecule. This study paves the way for further investigation of the role of Gal-9 in leukocyte recruitment in different inflammatory settings.
Sigma-Aldrich
Anti-CD18 Mouse mAb (IB4), liquid, ≥95% (SDS-PAGE), clone IB4
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid