Merck
  • Prosaposin maintains lipid homeostasis in dopamine neurons and counteracts experimental parkinsonism in rodents.
Prosaposin maintains lipid homeostasis in dopamine neurons and counteracts experimental parkinsonism in rodents.
Nature communications (2023-09-20)
Yachao He, Ibrahim Kaya, Reza Shariatgorji, Johan Lundkvist, Lars U Wahlberg, Anna Nilsson, Dejan Mamula, Jan Kehr, Justyna Zareba-Paslawska, Henrik Biverstål, Karima Chergui, Xiaoqun Zhang, Per E Andren, Per Svenningsson
ABSTRACT
Prosaposin (PSAP) modulates glycosphingolipid metabolism and variants have been linked to Parkinson's disease (PD). Here, we find altered PSAP levels in the plasma, CSF and post-mortem brain of PD patients. Altered plasma and CSF PSAP levels correlate with PD-related motor impairments. Dopaminergic PSAP-deficient (cPSAPDAT) mice display hypolocomotion and depression/anxiety-like symptoms with mildly impaired dopaminergic neurotransmission, while serotonergic PSAP-deficient (cPSAPSERT) mice behave normally. Spatial lipidomics revealed an accumulation of highly unsaturated and shortened lipids and reduction of sphingolipids throughout the brains of cPSAPDAT mice. The overexpression of α-synuclein via AAV lead to more severe dopaminergic degeneration and higher p-Ser129 α-synuclein levels in cPSAPDAT mice compared to WT mice. Overexpression of PSAP via AAV and encapsulated cell biodelivery protected against 6-OHDA and α-synuclein toxicity in wild-type rodents. Thus, these findings suggest PSAP may maintain dopaminergic lipid homeostasis, which is dysregulated in PD, and counteract experimental parkinsonism.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Tryptophan Hydroxylase antibody produced in mouse, clone WH-3, ascites fluid
Sigma-Aldrich
Anti-Chicken IgG (H+L), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Tamoxifen, ≥99%