Merck
  • Ginsenoside protopanaxadiol protects adult retinal pigment epithelial-19 cells from chloroquine by modulating autophagy and apoptosis.
Ginsenoside protopanaxadiol protects adult retinal pigment epithelial-19 cells from chloroquine by modulating autophagy and apoptosis.
PloS one (2022-12-02)
Haesung Lee, Anh Thu Nguyen Hoang, Sook-Jeong Lee
ABSTRACT
Chloroquine often causes serious eye and vision problems, which are mainly mediated by lysosomotropic alteration. In this study, we investigated whether the ginsenoside protopanaxadiol relieves chloroquine-induced retinopathy by restoring lysosomotropic abnormalities in human adult retinal pigment epithelial-19 cells. Cytotoxicity was assessed using a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay. Morphological alterations in autophagosomes of adult retinal pigment epithelial-19 cells was detected using confocal microscopy. Apoptosis was examined using flow cytometry, whereas cellular reactive oxygen species levels were determined by measuring the fluorescence intensity of 5-(and-6)-carboxy-2'-7'-dichlorohydrofluorescein diacetate. Lysosomal function was assessed by measuring lysosomal pH and enzyme activity. Immunoprecipitation and western blotting analyses were performed. Adult retinal pigment epithelial-19 cells accumulated autophagosomes with fusion defects in lysosomes and reactive oxygen species formation following exposure to chloroquine. This effect trapped Beclin-1 and B-cell lymphoma 2 interfering with autophagy initiation and autophagosome development. Protopanaxadiol alleviated chloroquine-induced toxicity by modulating the interaction between Beclin-1 and Bcl-2, which was mediated by the AMP-activated protein kinase-mammalian target of rapamycin signal axis. Furthermore, autophagy and apoptosis were simultaneously controlled by protopanaxadiol via upregulation of autophagy flux and decreased reactive oxygen species formation and apoptotic protein expression. These findings suggest that protopanaxadiol is a promising treatment strategy for chloroquine-mediated retinopathy.
Sigma-Aldrich
Staurosporine from Streptomyces sp., for molecular biology, ≥95% (HPLC)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Anti-TUBA4A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
ANTI-BECLIN1-BH3 DOMAIN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder