Merck
Deglycosylation of SLAMF7 in breast cancers enhances phagocytosis.
American journal of cancer research (2022-11-17)
Shih-Han Wang, Wen-Cheng Chou, Hsiang-Chi Huang, Te-An Lee, Tzu-Chun Hsiao, Ling-Hui Wang, Ke-Bin Huang, Chun-Tse Kuo, Chi-Hong Chao, Shing-Jyh Chang, Jung-Mao Hsu, Jialei Weng, Ning Ren, Fu-An Li, Yun-Ju Lai, Chenhao Zhou, Mien-Chie Hung, Chia-Wei Li
ABSTRACT
N-linked glycosylation of proteins is one of the post-translational modifications (PTMs) that shield tumor antigens from immune attack. Signaling lymphocytic activation molecule family 7 (SLAMF7) suppresses cancer cell phagocytosis and is an ideal target under clinical development. PTM of SLAMF7, however, remains less understood. In this study, we investigated the role of N-glycans on SLAMF7 in breast cancer progression. We identified seven N-linked glycosylation motifs on SLAMF7, which are majorly occupied by complex structures. Evolutionally conserved N98 residue is enriched with high mannose and sialylated glycans. Hyperglycosylated SLAMF7 was associated with STT3A expression in breast cancer cells. Inhibition of STT3A by a small molecule inhibitor, N-linked glycosylation inhibitor-1 (NGI-1), reduced glycosylation of SLAMF7, resulting in enhancing antibody affinity and phagocytosis. To provide an on-target effect, we developed an antibody-drug conjugate (ADC) by coupling the anti-SLAMF7 antibody with NGI-1. Deglycosylation of SLAMF7 increases antibody recognition and promotes macrophage engulfment of breast cancer cells. Our work suggests deglycosylation by ADC is a potential strategy to enhance the response of immunotherapeutic agents.
Sigma-Aldrich
NGI-1, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline