Merck
  • The Role of Interstitial Fluid Pressure in Cerebral Porous Biomaterial Integration.
The Role of Interstitial Fluid Pressure in Cerebral Porous Biomaterial Integration.
Brain sciences (2022-04-22)
Fabien Bonini, Sébastien Mosser, Flavio Maurizio Mor, Anissa Boutabla, Patrick Burch, Amélie Béduer, Adrien Roux, Thomas Braschler
ABSTRACT
Recent advances in biomaterials offer new possibilities for brain tissue reconstruction. Biocompatibility, provision of cell adhesion motives and mechanical properties are among the present main design criteria. We here propose a radically new and potentially major element determining biointegration of porous biomaterials: the favorable effect of interstitial fluid pressure (IFP). The force applied by the lymphatic system through the interstitial fluid pressure on biomaterial integration has mostly been neglected so far. We hypothesize it has the potential to force 3D biointegration of porous biomaterials. In this study, we develop a capillary hydrostatic device to apply controlled in vitro interstitial fluid pressure and study its effect during 3D tissue culture. We find that the IFP is a key player in porous biomaterial tissue integration, at physiological IFP levels, surpassing the known effect of cell adhesion motives. Spontaneous electrical activity indicates that the culture conditions are not harmful for the cells. Our work identifies interstitial fluid pressure at physiological negative values as a potential main driver for tissue integration into porous biomaterials. We anticipate that controlling the IFP level could narrow the gap between in vivo and in vitro and therefore decrease the need for animal screening in biomaterial design.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Collagen solution from bovine skin, Type I, 2.9-3.2 mg/mL, suitable for cell culture, sterile-filtered
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture