Merck
  • Rough-Form Lipopolysaccharide Increases Apoptosis in Human CD4⁺ and CD8⁺ T Lymphocytes.
Rough-Form Lipopolysaccharide Increases Apoptosis in Human CD4⁺ and CD8⁺ T Lymphocytes.
Scandinavian journal of immunology (2011-08-23)
J S Nielsen, A Larsson, T Ledet, M Turina, E Tønnesen, J Krog
ABSTRACT
Immunosuppression induced by lymphocyte apoptosis is considered an important factor in the pathogenesis of sepsis and has been demonstrated in both animal models of lipopolysaccharide (LPS)-induced endotoxemia and septic patients. As rough-form LPS (R-LPS) has recently been shown to elicit a stronger immunological response than regular smooth-form LPS (S-LPS), we aimed to assess the apoptosis-inducing capabilities of R-LPS in different subsets of lymphocytes (CD4(+) T cells, CD8(+) T cell, B cells and NK cells). Using multicolour flow cytometry on human peripheral blood mononuclear cells, we found that R-LPS increased apoptosis in CD4(+) and CD8(+) T cells assessed by annexin V and propidium iodide (AV(+) PI(-)), compared with both S-LPS-stimulated and unstimulated cells. 7-Amino-actinomycin D and staining for intracellular active caspase-3, which are considered later signs of apoptosis, did not reveal the same results. Both forms appeared to inhibit apoptosis in B cells, but no LPS-form-specific effect was seen on B or NK cells. Our results indicate that R-LPS induces a stronger AV(+) PI(-)-assessed apoptotic response in T cells than S-LPS. Our findings emphasize the importance of T cell apoptosis in endotoxemia and advocates for control of LPS form in both endotoxemia research and clinical trials with Gram-negative infections.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Escherichia coli F583 (Rd mutant)