Merck
  • Synthesis and substance P receptor binding activity of androstano[3,2-b]pyrimido[1,2-a]benzimidazoles.
Synthesis and substance P receptor binding activity of androstano[3,2-b]pyrimido[1,2-a]benzimidazoles.
Journal of medicinal chemistry (1992-01-24)
B R Venepalli, L D Aimone, K C Appell, M R Bell, J A Dority, R Goswami, P L Hall, V Kumar, K B Lawrence, M E Logan
ABSTRACT
Several heterosteroids containing a dihydroethisterone skeleton were prepared and shown to displace substance P in a receptor binding assay. Further biochemical (kinetic and Scatchard analyses) and pharmacological evaluation (substance P-induced plasma extravasation and salivation in the rat) of a representative example in this series (5a) established that these compounds are competitive antagonists at the substance P receptor.
Sigma-Aldrich
WIN 62,577, solid
Sigma-Aldrich
WIN 51708 hydrate, solid