Merck
  • Stem cell niche organization in the Drosophila ovary requires the ECM component Perlecan.
Stem cell niche organization in the Drosophila ovary requires the ECM component Perlecan.
Current biology : CB (2021-02-24)
Alfonsa Díaz-Torres, Alicia E Rosales-Nieves, John R Pearson, Carmen Santa-Cruz Mateos, Miriam Marín-Menguiano, Owen J Marshall, Andrea H Brand, Acaimo González-Reyes
ABSTRACT
Stem cells reside in specialized microenvironments or niches that balance stem cell proliferation and differentiation.1,2 The extracellular matrix (ECM) is an essential component of most niches, because it controls niche homeostasis, provides physical support, and conveys extracellular signals.3-11 Basement membranes (BMs) are thin ECM sheets that are constituted mainly by Laminins, Perlecan, Collagen IV, and Entactin/Nidogen and surround epithelia and other tissues.12 Perlecans are secreted proteoglycans that interact with ECM proteins, ligands, receptors, and growth factors such as FGF, PDGF, VEGF, Hedgehog, and Wingless.13-18 Thus, Perlecans have structural and signaling functions through the binding, storage, or sequestering of specific ligands. We have used the Drosophila ovary to assess the importance of Perlecan in the functioning of a stem cell niche. Ovarioles in the adult ovary are enveloped by an ECM sheath and possess a tapered structure at their anterior apex termed the germarium. The anterior tip of the germarium hosts the germline niche, where two to four germline stem cells (GSCs) reside together with a few somatic cells: terminal filament cells (TFCs), cap cells (CpCs), and escort cells (ECs).19 We report that niche architecture in the developing gonad requires trol, that niche cells secrete an isoform-specific Perlecan-rich interstitial matrix, and that DE-cadherin-dependent stem cell-niche adhesion necessitates trol. Hence, we provide evidence to support a structural role for Perlecan in germline niche establishment during larval stages and in the maintenance of a normal pool of stem cells in the adult niche.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Roche
RNase, DNase-free, from bovine pancreas
Sigma-Aldrich
Trizma® hydrochloride solution, pH 8.0, BioPerformance Certified, 1 M, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Phosphate buffered saline, tablet