Merck
  • Integrated multi-wavelength microscope combining TIRFM and IRM modalities for imaging cellulases and other processive enzymes.
Integrated multi-wavelength microscope combining TIRFM and IRM modalities for imaging cellulases and other processive enzymes.
Biomedical optics express (2021-07-06)
Daguan Nong, Zachary K Haviland, Kate Vasquez Kuntz, Ming Tien, Charles T Anderson, William O Hancock
ABSTRACT
We describe a multimodal microscope for visualizing processive enzymes moving on immobilized substrates. The instrument combines interference reflection microscopy (IRM) with multi-wavelength total internal reflectance fluorescence microscopy (TIRFM). The microscope can localize quantum dots with a precision of 2.8 nm at 100 frames/s, and was used to image the dynamics of the cellulase, Cel7a interacting with surface-immobilized cellulose. The instrument, which was built with off-the-shelf components and is controlled by custom software, is suitable for tracking other degradative enzymes such as collagenases, as well as motor proteins moving along immobilized tracks.
Sigma-Aldrich
Cellobiohydrolase I from Hypocrea jecorina, 0.13 U/mg, recombinant, expressed in corn