Merck
  • Expression of C9orf72 hexanucleotide repeat expansion leads to formation of RNA foci and dipeptide repeat proteins but does not influence autophagy or proteasomal function in neuronal cells.
Expression of C9orf72 hexanucleotide repeat expansion leads to formation of RNA foci and dipeptide repeat proteins but does not influence autophagy or proteasomal function in neuronal cells.
Biochimica et biophysica acta. Molecular cell research (2021-03-30)
Stina Leskelä, Nadine Huber, Dorit Hoffmann, Hannah Rostalski, Anne M Remes, Mari Takalo, Mikko Hiltunen, Annakaisa Haapasalo
ABSTRACT
C9orf72 hexanucleotide repeat expansion (HRE) is the major genetic cause underpinning frontotemporal lobar degeneration (FLTD) and amyotrophic lateral sclerosis (ALS). C9orf72 HRE-associated pathogenesis involves both loss-of-function, through reduced C9orf72 levels, and gain-of-function mechanisms, including formation of RNA foci and generation of dipeptide repeat (DPR) proteins. In addition, dysfunctional protein degradation pathways, i.e. autophagy and ubiquitin-proteasome system (UPS), are suggested. Our aim was to study the gain-of-function mechanisms in the context of the function of protein degradation pathways as well as the regulation of the DPR proteins through these pathways. To this end, we expressed the pathological HRE in neuronal N2a cells and mouse primary cortical neurons. Protein degradation pathways were modulated to induce or block autophagy or to inhibit UPS. In addition, proteasomal activity was assessed. The C9orf72 HRE-expressing N2a cells and neurons were confirmed to produce RNA foci and DPR proteins, predominantly the Poly-GP proteins. However, the presence of these pathological hallmarks did not result in alterations in autophagy or proteasomal activity in either of the studied cell types. In N2a cells, Poly-GP proteins appeared in soluble forms and Lactacystin-mediated UPS inhibition increased their levels, indicating proteasomal regulation. Similar effects were not observed in cortical neurons, where the Poly-GP proteins formed also higher molecular weight forms. These results suggest a cell type-specific morphology and regulation of the DPR proteins. Further studies in other model systems may shed additional light onto the effects of the C9orf72 HRE on cellular protein degradation pathways and the regulation of the DPR protein levels.
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GR) Antibody, clone 5A2, clone 5A2, 1 mg/mL, from rat
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-PA), serum, from rabbit
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GA) Antibody, clone 5E9, clone 5E9, from mouse