Merck
  • Anion inhibition studies of an α-carbonic anhydrase from the living fossil Astrosclera willeyana.
Anion inhibition studies of an α-carbonic anhydrase from the living fossil Astrosclera willeyana.
Bioorganic & medicinal chemistry letters (2012-01-10)
Anna Ohradanova, Daniela Vullo, Silvia Pastorekova, Jaromir Pastorek, Daniel J Jackson, Gert Wörheide, Claudiu T Supuran
ABSTRACT
An α-carbonic anhydrase (CA, EC 4.2.1.1) isolated from the living fossil sponge Astrosclera willeyana, Astrosclerin, was investigated for its inhibition profile with simple inorganic anions, complex anions and other small molecules known to interact with these zinc enzymes. Astrosclerin is a catalytically highly efficient enzyme, and is inhibited in the low micromolar range by sulfamide, sulfamic acid, phenylboronic acid and phenylarsonic acid, and in the submillimolar range by a variety of anions including fluoride, chloride, cyanate, thiocyanate, cyanide, hydrogen sulfide, bisulfate, stannate, perosmate, divanadate, perrhenate, perruthenate, selenocyanide, trithiocarbonate, diethyldithiocarbamate and iminodisulfonate. Less efficient Astrosclerin inhibitors were sulfate, bromide, iodide, azide, bicarbonate, carbonate, tetraborate and perchlorate (K(I)s of 5.11-30.6mM) whereas tetrafluoroborate was not at all inhibitory. Because Astrosclerin is involved in calcification processes in vivo, its anion inhibition profile may be important for future studies designed to shed light on the physiologic functions of α-CAs in marine organisms.
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium chloride physiological solution, BioUltra, tablet
Sigma-Aldrich
Sodium chloride physiological solution, BioUltra, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium nitrite solution
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%