- High throughput deep sequencing elucidates the important role of lncRNAs in Foxtail millet response to herbicides.
High throughput deep sequencing elucidates the important role of lncRNAs in Foxtail millet response to herbicides.
Long non-coding RNAs (lncRNAs) play an important function in plant growth and development as well as response to stresses. However, little information was known in foxtail millet; no study was reported on lncRNAs in plant response to herbicide treatment. In this study, by using deep sequencing and advanced bioinformatic analysis, a total of 2547 lncRNAs were identified, including 787 known and 1760 novel lncRNAs. These lncRNAs are distributed across all 9 chromosomes, and the majority were located in the intergenic region with 1-2 exons. These lncRNAs were differentially expressed between different genotypes under different herbicide treatments. lncRNAs regulate plant growth and development as well as response to herbicide treatments through targeting protein-coding genes that directly relate to chemical metabolism and defense system. Multiple potential target genes and lncRNA-mRNA-miRNA gene networks were discovered. These results elucidate the potential roles of lncRNAs in plant response to herbicides.