Merck
  • Calibration of organic-diffusive gradients in thin films (o-DGT) passive samplers for perfluorinated alkyl acids in water.
Calibration of organic-diffusive gradients in thin films (o-DGT) passive samplers for perfluorinated alkyl acids in water.
Chemosphere (2020-12-11)
Po Wang, Jonathan K Challis, Kim H Luong, Trisha C Vera, Charles S Wong
ABSTRACT
The application of the organic-diffusive gradients in thin films (o-DGT) passive sampling technique for the monitoring of per- and polyfluoroalkyl substances (PFAS) in the environment is still limited. Six common PFAS with different chain lengths were evaluated in water by o-DGT. Measured diffusion coefficients (D) in agarose and polyacrylamide diffusive gels ranged from 4.55-8.63 × 10-6 cm2 s-1 and 3.85-7.00 × 10-6 cm2 s-1 at 23 °C, respectively. Experimental sampling rates (Rs) for both agarose- and polyacrylamide-WAX sampler configurations were within 22% relative error of D-based Rs for four of the PFAS. Larger differences for perfluorobutanesulfonic acid (PFBS) and perfluoroundecanoic acid (PFUnDA) ranged from 36% to 56%. In general, in-situ Rs can be predicted using measured D-values for perfluorinated alkyl acids. The mass accumulation of six PFAS in two o-DGT configurations was linear over 21 days (R2 ≥ 0.97). Diffusion and uptake of o-DGT depended on the gel type and specific PFAS. Field demonstrations of o-DGT with WAX and HLB binding gels and polyacrylamide diffusive gels (not prone to biodegradation) found 0.3-19.5 ng L-1 of PFAS in rivers near industrial areas around Guangzhou and Foshan, China, with no apparent differences between the two co-deployed samplers. This study demonstrates that the configurations of o-DGT tested provide a cost-effective monitoring tool for measuring perfluorinated alkyl acids in aquatic systems, in particular the four PFAS for which reasonable correlations were observed.
Sigma-Aldrich
Perfluorodecanoic acid, 98%
Sigma-Aldrich
Perfluoroundecanoic acid, 95%
Sigma-Aldrich
Nonafluorobutane-1-sulfonic acid, 97%
Sigma-Aldrich
Perfluorononanoic acid, 97%