Merck
  • BPRDP056, a novel small molecule drug conjugate specifically targeting phosphatidylserine for cancer therapy.
BPRDP056, a novel small molecule drug conjugate specifically targeting phosphatidylserine for cancer therapy.
Translational oncology (2020-10-18)
Yun-Yu Chen, Chen-Fu Lo, Tai-Yu Chiu, Chia-Yu Hsu, Teng-Kuang Yeh, Ching-Ping Chen, Chen-Lung Huang, Chung-Yu Huang, Min-Hsien Wang, Yu-Chen Huang, Hsuan-Hui Ho, Yu-Sheng Chao, Joe C Shih, Lun K Tsou, Chiung-Tong Chen
ABSTRACT
Zinc(II)-dipicolylamine (Zn-DPA) has been shown to specifically identify and bind to phosphatidylserine (PS), which exists in bulk in the tumor microenvironment. BPRDP056, a Zn-DPA-SN38 conjugate was designed to provide PS-targeted drug delivery of a cytotoxic SN38 to the tumor microenvironment, thereby allowing a lower dosage of SN38 that induces apoptosis in cancer cells. Micro-Western assay showed that BPRDP056 exhibited apoptotic signal levels similar to those of CPT-11 in the treated tumors growing in mice. Pharmacokinetic study showed that BPRDP056 has excellent systemic stability in circulation in mice and rats. BPRDP056 is accumulated in tumors and thus increases the cytotoxic effects of SN38. The in vivo antitumor activities of BPRDP056 have been shown to be significant in subcutaneous pancreas, prostate, colon, liver, breast, and glioblastoma tumors, included an orthotopic pancreatic tumor, in mice. BPRDP056 shrunk tumors at a lower (~20% only) dosing intensity of SN38 compared to that of SN38 conjugated in CPT-11 in all tumor models tested. A wide spectrum of antitumor activities is expected to treat all cancer types of PS-rich tumor microenvironments. BPRDP056 is a first-in-class small molecule drug conjugate for cancer therapy.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Supelco
Temozolomide, Pharmaceutical Secondary Standard; Certified Reference Material