Merck
PLA micro- and nano-particles.
Advanced drug delivery reviews (2016-06-06)
Byung Kook Lee, Yeonhee Yun, Kinam Park
ABSTRACT
Poly(d,l-lactic acid) (PLA) has been widely used for various biomedical applications for its biodegradable, biocompatible, and nontoxic properties. Various methods, such as emulsion, salting out, and precipitation, have been used to make better PLA micro- and nano-particle formulations. They are widely used as controlled drug delivery systems of therapeutic molecules, including proteins, genes, vaccines, and anticancer drugs. Even though PLA-based particles have challenges to overcome, such as low drug loading capacity, low encapsulation efficiency, and terminal sterilization, continuous innovations in particulate formulations will lead to development of clinically useful formulations.
Sigma-Aldrich
NanoFabTx materials screening kit, for synthesis of polymeric nanoparticles
Sigma-Aldrich
NanoFabTx PEGylated nanoparticle formulation screening kit
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PCL drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PLA-nano, for synthesis of 100 and 200 nm particles
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PLA drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PEG-PLA drug formulation screening kit, for synthesis of PEGylated nanoparticles