Merck
A Neural Network for Wind-Guided Compass Navigation.
Neuron (2020-07-19)
Tatsuo S Okubo, Paola Patella, Isabel D'Alessandro, Rachel I Wilson
ABSTRACT
Spatial maps in the brain are most accurate when they are linked to external sensory cues. Here, we show that the compass in the Drosophila brain is linked to the direction of the wind. Shifting the wind rightward rotates the compass as if the fly were turning leftward, and vice versa. We describe the mechanisms of several computations that integrate wind information into the compass. First, an intensity-invariant representation of wind direction is computed by comparing left-right mechanosensory signals. Then, signals are reformatted to reduce the coding biases inherent in peripheral mechanics, and wind cues are brought into the same circular coordinate system that represents visual cues and self-motion signals. Because the compass incorporates both mechanosensory and visual cues, it should enable navigation under conditions where no single cue is consistently reliable. These results show how local sensory signals can be transformed into a global, multimodal, abstract representation of space.
Product Description
Supelco
Liquid scintillation vials, glass, volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with Poly-Seal cone liner. (caps packed separately), pkg of 500 ea
Pricing and availability is not currently available.
Sigma-Aldrich
Picrotoxin, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Goat serum
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, for molecular biology
Pricing and availability is not currently available.