Merck
  • Single-Cell Profiling Reveals Sex, Lineage, and Regional Diversity in the Mouse Kidney.
Single-Cell Profiling Reveals Sex, Lineage, and Regional Diversity in the Mouse Kidney.
Developmental cell (2019-11-07)
Andrew Ransick, Nils O Lindström, Jing Liu, Qin Zhu, Jin-Jin Guo, Gregory F Alvarado, Albert D Kim, Hannah G Black, Junhyong Kim, Andrew P McMahon
ABSTRACT
Chronic kidney disease affects 10% of the population with notable differences in ethnic and sex-related susceptibility to kidney injury and disease. Kidney dysfunction leads to significant morbidity and mortality and chronic disease in other organ systems. A mouse-organ-centered understanding underlies rapid progress in human disease modeling and cellular approaches to repair damaged systems. To enhance an understanding of the mammalian kidney, we combined anatomy-guided single-cell RNA sequencing of the adult male and female mouse kidney with in situ expression studies and cell lineage tracing. These studies reveal cell diversity and marked sex differences, distinct organization and cell composition of nephrons dependent on the time of nephron specification, and lineage convergence, in which contiguous functionally related cell types are specified from nephron and collecting system progenitor populations. A searchable database, Kidney Cell Explorer (https://cello.shinyapps.io/kidneycellexplorer/), enables gene-cell relationships to be viewed in the anatomical framework of the kidney.
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Anti-SLC26A4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid