Merck
Subdiffraction resolution in far-field fluorescence microscopy.
Optics letters (2007-12-13)
T A Klar, S W Hell
ABSTRACT
We overcame the resolution limit of scanning far-field fluorescence microscopy by disabling the fluorescence from the outer part of the focal spot. Whereas a near-UV pulse generates a diffraction-limited distribution of excited molecules, a spatially offset pulse quenches the excited molecules from the outer part of the focus through stimulated emission. This results in a subdiffraction-sized effective point-spread function. For a 1.4 aperture and a 388-nm excitation wavelength spatial resolution is increased from 150 +/- 8 nm to 106 +/- 8 nm with a single offset beam. Superior lateral resolution is demonstrated by separation of adjacent Pyridine 2 nanocrystals that are otherwise indiscernible.
Sigma-Aldrich
Abberior® STAR 440SXP, maleimide, for long Stokes STED and 2-color STED application
Sigma-Aldrich
Abberior® STAR 440SXP, NHS ester, for long Stokes STED and 2-color STED application
Sigma-Aldrich
Abberior® STAR 580, NHS ester, for STED application
Sigma-Aldrich
Abberior® STAR 512, maleimide, for STED application
Sigma-Aldrich
Abberior® STAR 635, NHS ester, for STED application
Sigma-Aldrich
Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)