Merck
  • All-Electrical Ca2+-Independent Signal Transduction Mediates Attractive Sodium Taste in Taste Buds.
All-Electrical Ca2+-Independent Signal Transduction Mediates Attractive Sodium Taste in Taste Buds.
Neuron (2020-04-02)
Kengo Nomura, Miho Nakanishi, Fumiyoshi Ishidate, Kazumi Iwata, Akiyuki Taruno
ABSTRACT
Sodium taste regulates salt intake. The amiloride-sensitive epithelial sodium channel (ENaC) is the Na+ sensor in taste cells mediating attraction to sodium salts. However, cells and intracellular signaling underlying sodium taste in taste buds remain long-standing enigmas. Here, we show that a subset of taste cells with ENaC activity fire action potentials in response to ENaC-mediated Na+ influx without changing the intracellular Ca2+ concentration and form a channel synapse with afferent neurons involving the voltage-gated neurotransmitter-release channel composed of calcium homeostasis modulator 1 (CALHM1) and CALHM3 (CALHM1/3). Genetic elimination of ENaC in CALHM1-expressing cells as well as global CALHM3 deletion abolished amiloride-sensitive neural responses and attenuated behavioral attraction to NaCl. Together, sodium taste is mediated by cells expressing ENaC and CALHM1/3, where oral Na+ entry elicits suprathreshold depolarization for action potentials driving voltage-dependent neurotransmission via the channel synapse. Thus, all steps in sodium taste signaling are voltage driven and independent of Ca2+ signals. This work also reveals ENaC-independent salt attraction.
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Donkey Anti-Guinea Pig IgG Antibody, biotin conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Carbenoxolone disodium salt, ≥98%
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Barium chloride dihydrate, ≥99%