Merck
  • Neuroprotective Effects of Direct Intrathecal Administration of Granulocyte Colony-Stimulating Factor in Rats with Spinal Cord Injury.
Neuroprotective Effects of Direct Intrathecal Administration of Granulocyte Colony-Stimulating Factor in Rats with Spinal Cord Injury.
CNS neuroscience & therapeutics (2015-07-21)
Wu-Fu Chen, Chun-Hong Chen, Nan-Fu Chen, Chun-Sung Sung, Zhi-Hong Wen
ABSTRACT
To date, no reliable methods have proven effective for treating spinal cord injury (SCI). Even systemic administration of methylprednisolone (MP) remains controversial. We previously reported that intrathecal (i.t.) administration of granulocyte colony-stimulating factor (G-CSF) improves outcome after experimental spinal cord ischemic insults in rats. The present study aimed to examine the neuroprotective efficacy of i.t. G-CSF or MP in rats with SCI. Female rats were subjected to spinal cord contusion injury at T10 using NYU impactor. We i.t. administered G-CSF (10 μg) or MP (one bolus of 100 μg, followed by 18 μg/h infusion for 23 h) immediately after SCI. Both G-CSF and MP significantly improved the rats' motor function after SCI. Immunofluorescence staining revealed suppressed expression of transforming growth factor-beta 1 (TGF-β1), chondroitin sulfate proteoglycans (neurocan and phosphacan), OX-42 and tumor necrosis factor alpha after i.t. G-CSF, but not MP, in rats with SCI. In addition, G-CSF significantly decreased the expression of astrocytic TGF-β1 and glial fibrillary acidic protein around the injury site. Furthermore, rats with G-CSF treatment showed increased neurofilament expression beyond the glial scars. Direct i.t. administration of G-CSF provides a promising therapeutic option for SCI or related spinal diseases.
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone RT97, clone RT97, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurocan Antibody, clone 650.24, clone 650.24, Chemicon®, from mouse
Sigma-Aldrich
Anti-Phosphacan Antibody, clone 122.2, ascites fluid, clone 122.2, Chemicon®