Merck
  • Characterization of rac and cdc42 activation in chemoattractant-stimulated human neutrophils using a novel assay for active GTPases.
Characterization of rac and cdc42 activation in chemoattractant-stimulated human neutrophils using a novel assay for active GTPases.
The Journal of biological chemistry (1999-05-01)
V Benard, B P Bohl, G M Bokoch
ABSTRACT
A major function of Rac2 in neutrophils is the regulation of oxidant production important in bacterial killing. Rac and the related GTPase Cdc42 also regulate the dynamics of the actin cytoskeleton, necessary for leukocyte chemotaxis and phagocytosis of microorganisms. Although these GTPases appear to be critical downstream components of chemoattractant receptor signaling in human neutrophils, the pathways involved in direct control of Rac/Cdc42 activation remain to be determined. We describe an assay that measures the formation of Rac-GTP and Cdc42-GTP based on their specific binding to the p21-binding domain of p21-activated kinase 1. A p21-binding domain glutathione S-transferase fusion protein specifically binds Rac and Cdc42 in their GTP-bound forms both in vitro and in cell samples. Binding is selective for Rac and Cdc42 versus RhoA. Using this assay, we investigated Rac and Cdc42 activation in neutrophils and differentiated HL-60 cells. The chemoattractant fMet-Leu-Phe and the phorbol ester phorbol myristate acetate stimulate formation of Rac-GTP and Cdc42-GTP with distinct time courses that parallel cell activation. We also show that the signaling pathways leading to Rac and Cdc42 activation in HL-60 cells involve G proteins sensitive to pertussis toxin, as well as tyrosine kinase and phosphatidylinositol 3-kinase activities.
Sigma-Aldrich
PAK-1 PBD Protein (agarose free), 300 µg, For use in Affinity Binding Assays, ELISA & G-Protein Assays.
Sigma-Aldrich
Cdc42 Activation Assay Kit, Non-radioactive cdc42 Activation Assay Kit can be used to precipitate cdc42-GTP from cell lysates subjected to various treatments & detection by a cdc42 specific monoclonal antibody.
Sigma-Aldrich
Rac1 Activation Assay Kit, Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates & detection by a Rac1 specific monoclonal antibody.
Sigma-Aldrich
Rac/cdc42 Assay Reagent (PAK1 PBD, agarose), 300 µg, Specifically binds to & precipitates Rac-GTP & cdc42-GTP from cell lysates. For use in Affinity Binding Assays.