Merck
  • Fluorinated Analogues of Desferrioxamine B from Precursor-Directed Biosynthesis Provide New Insight into the Capacity of DesBCD.
Fluorinated Analogues of Desferrioxamine B from Precursor-Directed Biosynthesis Provide New Insight into the Capacity of DesBCD.
ACS chemical biology (2018-08-08)
Thomas J Telfer, Rachel Codd
ABSTRACT
The siderophore desferrioxamine B (DFOB, 1) native to Streptomyces pilosus is biosynthesized by the DesABCD enzyme cluster. DesA-mediated decarboxylation of l-lysine gives 1,5-diaminopentane (DP) for processing by DesBCD. S. pilosus culture medium was supplemented with rac-1,4-diamino-2-fluorobutane ( rac-FDB) to compete against DP to generate fluorinated analogues of DFOB, as agents of potential clinical interest. LC-MS/MS analysis identified fluorinated analogues of DFOB with one, two, or three DP units (binary notation: 0) exchanged for one (DFOA-F1[001] (2), DFOA-F1[010] (3), DFOA-F1[100] (4)), two (DFOA-F2[011] (5), DFOA-F2[110] (6), DFOA-F2[101] (7)), or three (DFOA-F3[111] (8)) rac-FDB units (binary notation: 1). The two sets of constitutional isomers 2-4 and 5-7 arose from the position of the substrates in the N-acetyl, internal, or amine-containing regions of the DFOB trimer. N-Acetylated fluorinated DFOB analogues were formed where the rac-FDB substrate was positioned in the amine region ( e.g., N-Ac-DFOA-F1[001] (2a)). Other analogues contained two hydroxamic acid groups and three amide bonds. Experiments using rac-FDB, R-FDB, or S-FDB showed a similar species profile between rac-FDB and R-FDB. These data are consistent with the following. (i) DesB can act on rac-FDB. (ii) DesC can act directly on rac-FDB. (iii) The products of DesBC or DesC catalysis of rac-FDB can undergo a second round of DesC catalysis at the free amine. (iv) DesD catalysis of these products gives N, N'-diacetylated compounds. (v) A minimum of two hydroxamic acid groups is required to form a viable DesD-substrate(s) precomplex. (vi) One or more DesBCD-catalyzed steps in DFOB biosynthesis is enantioselective. This work has provided a potential path to access fluorinated analogues of DFOB and new insight into its biosynthesis.
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Chromeazurol S, Dye content 45 %