Merck
  • Antimelanogenesis and skin-protective activities of Panax ginseng calyx ethanol extract.
Antimelanogenesis and skin-protective activities of Panax ginseng calyx ethanol extract.
Journal of ginseng research (2018-07-10)
Jeong-Ook Lee, Eunji Kim, Ji Hye Kim, Yo Han Hong, Han Gyung Kim, Deok Jeong, Juewon Kim, Su Hwan Kim, Chanwoong Park, Dae Bang Seo, Young-Jin Son, Sang Yun Han, Jae Youl Cho
ABSTRACT
The antioxidant effects of Panax ginseng have been reported in several articles; however, little is known about the antimelanogenesis effect, skin-protective effect, and cellular mechanism of Panax ginseng, especially of P. ginseng calyx. To understand how an ethanol extract of P. ginseng berry calyx (Pg-C-EE) exerts skin-protective effects, we studied its activities in activated melanocytes and reactive oxygen species (ROS)-induced keratinocytes. To confirm the antimelanogenesis effect of Pg-C-EE, we analyzed melanin synthesis and secretion and messenger RNA and protein expression levels of related genes. Ultraviolet B (UVB) and hydrogen peroxide (H2O2) were used to induce cell damage by ROS generation. To examine whether this damage is inhibited by Pg-C-EE, we performed cell viability assays and gene expression and transcriptional activation analyses. Pg-C-EE inhibited melanin synthesis and secretion by blocking activator protein 1 regulatory enzymes such as p38, extracellular signal-regulated kinases (ERKs), and cyclic adenosine monophosphate response element-binding protein. Pg-C-EE also suppressed ROS generation induced by H2O2 and UVB. Treatment with Pg-C-EE decreased the expression of matrix metalloproteinases, mitogen-activated protein kinases, and hyaluronidases and increased the cell survival rate. These results suggest that Pg-C-EE may have antimelanogenesis properties and skin-protective properties through regulation of activator protein 1 and cyclic adenosine monophosphate response element-binding protein signaling. Pg-C-EE may be used as a skin-improving agent, with moisture retention and whitening effects.
Sigma-Aldrich
L-Dopa-(phenyl-d3), ≥98 atom % D, ≥98% (CP)
Pricing and availability is not currently available.