Merck
  • Correction to: Antibiotic resistance pattern and virulence genes content in avian pathogenic Escherichia coli (APEC) from broiler chickens in Chitwan, Nepal.
Correction to: Antibiotic resistance pattern and virulence genes content in avian pathogenic Escherichia coli (APEC) from broiler chickens in Chitwan, Nepal.
BMC veterinary research (2018-05-24)
Manita Subedi, Rebanta Kumar Bhattarai, Bhuminand Devkota, Sarita Phuyal, Himal Luitel
ABSTRACT
The original article [1] contains errors in author panels and their contributions, errors in both the Methodology and the Results sections, and errors with respect to funding sources. The affected sections of the manuscript and their respective regions of corrected text can be viewed ahead.
Sigma-Aldrich
Agarose