
Zearalenone

Synonym(s):
F-2 toxin
Empirical Formula (Hill Notation):
C18H22O5
CAS Number:
Molecular Weight:
318.36
Products
Product Category
Brand
Formula Weight
Gene Alias
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing