
Tetrakis(diethylamido)hafnium(IV)

Synonym(s):
Tetrakis(diethylamino)hafnium(IV)
Linear Formula:
[(CH2CH3)2N]4Hf
CAS Number:
Molecular Weight:
467.01
Products
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing