
Tetrakis(4-aminophenyl)methane

Empirical Formula (Hill Notation):
C25H24N4
CAS Number:
Molecular Weight:
380.48
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing