
Sumatriptan

Synonym(s):
3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide, Sumatriptan
Empirical Formula (Hill Notation):
C14H21N3O2S
CAS Number:
Molecular Weight:
295.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing