
Stanozolol

Synonym(s):
17β-Hydroxy-17α-methyl-5α-androstano[3,2-c]pyrazole, 5α-Androstane-17α-methyl-17β-ol-[3,2-c]pyrazole, Androstanazole, Stanazol
Empirical Formula (Hill Notation):
C21H32N2O
CAS Number:
Molecular Weight:
328.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing