Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium hexametaphosphate
Synonym(s):
Calgon, Phosphate glass, water soluble, Polyphosphate sodium salt, Sodium polyphosphate
CAS Number:
68915-31-1
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Physical Form
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
305553
Sodium hexametaphosphate,
crystalline, +200 mesh, 96%
Expand
View Pricing
71600
Sodium hexametaphosphate,
65-70% P
2
O
5
basis
Expand
View Pricing
28-2880
Sodium hexametaphosphate,
SAJ first grade, 65.0-83.0% P
2
O
5
basis
Expand
View Pricing