![(S)-Mevalonic acid lithium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/217/137/8c17fd5d-fc1f-4910-9ffb-f539fb3bd93b/640/8c17fd5d-fc1f-4910-9ffb-f539fb3bd93b.png)
(S)-Mevalonic acid lithium salt
![(S)-Mevalonic acid lithium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/217/137/8c17fd5d-fc1f-4910-9ffb-f539fb3bd93b/640/8c17fd5d-fc1f-4910-9ffb-f539fb3bd93b.png)
Synonym(s):
Lithium (S)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (S)-3,5-dihydroxy-3-methyl-valerate, S-MVA-Li
Empirical Formula (Hill Notation):
C6H12O4 · xLi+
CAS Number:
Molecular Weight:
148.16 (free acid basis)
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing